Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™

Catalog No. MP219484805 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg

Used in a wide variety of biochemical applications. Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals is an essential nutrient found widely in nature. It is used as a buffer component, in precipitation and chemical sequencing of DNA, in chr

Sodium chloride is used in a wide variety of biochemical applications, including:

  • Intravenous fluids (normal saline is 0.9% w/v in water10)
  • Density gradients
  • A diluent to increase ionic strength in buffers or culture media
  • In salt-out procedures in the isolation of DNA
  • In high concentrations for preservation of foods, since bacteria cannot grow in high salt conditions

A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as &minus’21°C, depending on the rate of stirring and the size of the ice chunks. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Physical Form Crystalline Powder
Quantity 5 kg
Solubility Information Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl.
IUPAC Name sodium chloride
Formula Weight 58.44g/mol
Percent Purity ≥99.5%
Grade Molecular Biology
Recommended Storage Store at Room Temperature (15–30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.