Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloride, Optical Grade

Catalog No. AA1101930
Click to view available options
Quantity:
50 g
250 g

CAS: 7647-14-5 | ClNa | 58.44 g/mol

Sodium Chloride (NaCl) is widely used in biochemistry and molecular biology applications. It is considered an essential nutrient. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. NaCl is a component of phosphate buffered saline and SSC buffer. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
Color White
pH 6 to 9
Physical Form Broken Crystals
Quantity 250 g
Merck Index 14,8599
Solubility Information Soluble in water at 20°C 358g/L. Very slightly soluble in alcohol,glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid.
Formula Weight 58.44
Grade Optical
Odor Odorless
Refractive Index 1.5442
Sensitivity Hygroscopic
Density 2.165 g/mL
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3
RTECSNumber VZ4725000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.