Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Saturated (L-012), Reagents

Catalog No. 01333651
Change view
Click to view available options
:
Quantity: 10 L
Catalog No.
01-333-651 Quantity: 10 L
1 options

Catalog No. 01-333-651

Supplier: Reagents Holdings Llc CS14200010F

Only null left
Add to Cart
Add to Cart

Sodium Chloride, Saturated, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Table salt solution
IUPAC Name water
SMILES O

Specifications

Boiling Point 103°C
Color Colorless
Physical Form Liquid
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.202
Chemical Name or Material Sodium Chloride, Saturated (L-012)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.