Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chloride Solution 700 g NaCl and 4000 g H2O Ratio, Reagents
Click to view available options
:
Quantity: 10 L
Specifications
Specifications
Chemical Name or Material | Sodium Chloride Solution |
Boiling Point | 103°C |
CAS | 7732-18-5, 7647-14-5 |
Color | Colorless |
pH | 5 to 9 |
Physical Form | Liquid |
Quantity | 10 L |
Molecular Formula | NaCl/H2O |
Synonym | Table salt solution |
Grade | Reagent |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction