Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chlorite, tech. nominally 80%

Catalog No. p-4407876
Click to view available options
Quantity:
500 g
2 kg
10 kg

Sodium chlorite, ClNaO3, CAS Number-7758-19-2, 7775-09-9, 1310-73-2, 497-19-8, 7647-14-5, 7757-82-6, chlorate de sodium, asex, chloric acid, sodium salt, atlacide, tumbleleaf, rasikal, kusatol, chlorsaure, desolet, sodium chlorate, 10kg, White flake/powder, Nominally 80%, CHEBI:65242 | CAS: 7758-19-2 | ClNaO3 | 106.44 g/mol

Sodium chlorite is used in the in situ generation of chlorine dioxide for stripping of textiles, bleaching, pulp and paper industries. It acts as a disinfectant in water treatment plant and as a preservative in eye drops. It is also used in contact lens cleaning solution and for sanitizing air ducts. It is associated with zinc chloride and used as a component in therapeutic rinses, toothpastes, mouth sprays and gels. It is utilized for the synthesis of 4-oxo-2-alkenoic acids from alkyl furans. Further, it is involved as a reagent in Pinnick oxidation reaction to prepare carboxylic acid from aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2, 7775-09-9, 1310-73-2, 497-19-8, 7647-14-5, 7757-82-6
MDL Number MFCD00003478
IUPAC Name sodium;chlorite

Specifications

Melting Point 180°C to 200°C (decomposition)
Color White
Physical Form Flakes or Powder
Quantity 500 g
Assay Percent Range Nominally 80%
Molecular Formula ClNaO3
Synonym sodium;chlorite, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
Solubility Information Soluble in water. Slightly soluble in methanol and ethanol.
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
SMILES [Na+].[O-][Cl](=O)=O
Molecular Weight (g/mol) 106.44
PubChem CID 516902
ChEBI CHEBI:65242
Formula Weight 90.44
Grade Technical
Concentration or Composition (by Analyte or Components) Sodium chlorite: 80.0%; Sodium chlorate: 3.0%; Sodium hydroxide: 3.0%; Sodium carbonate: 2.0%; Sodium chloride: 10.0%; Sodium sulphate: 0.3%; Water: 1.7%
Sensitivity Hygroscopic
Appearance White flake/powder
Chemical Name or Material Sodium chlorite
Show More Show Less
Hazard Category H271-H301-H310-H314-H373
Precautionary Statement P210-P220-P221-P260-P262-P264b-P270-P271-P280-P283-P284-P302+P350-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1, Packing Group: II
EINECSNumber 231-836-6
RTECSNumber VZ4800000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.