Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloroacetate, 98%

Catalog No. AAA1237936
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g

CAS: 3926-62-3 | C2H2ClNaO2 | 116.476 g/mol

Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

Solubility
Soluble in water, ether, chloroform, benzene and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3926-62-3
Molecular Formula C2H2ClNaO2
Molecular Weight (g/mol) 116.476
MDL Number MFCD00002684
InChI Key FDRCDNZGSXJAFP-UHFFFAOYSA-M
Synonym sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech
PubChem CID 23665759
IUPAC Name sodium;2-chloroacetate
SMILES C(C(=O)[O-])Cl.[Na+]

Specifications

Melting Point ∽199°C (decomposition)
Flash Point 270°C (518°F)
Assay Percent Range 98%
Linear Formula CH2CO2Na
Quantity 500 g
UN Number UN2659
Beilstein 3597157
Merck Index 14,2112
Solubility Information Soluble in water,ether,chloroform,benzene and alcohol.
Formula Weight 116.48
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium Chloroacetate
Show More Show Less
Hazard Category H301-H315
Hazard Statement GHS H Statement
H301-H315
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM CHLOROACETATE
EINECSNumber 223-498-3
RTECSNumber AG1400000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.