Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium chromate tetrahydrate, 99+%

Click to view available options
Quantity:
100g
500g
10034-82-9
CrH8Na2O8
234.032
MFCD00149167
AVHUVTPOXZEIRG-UHFFFAOYSA-N
sodium chromate tetrahydrate,cro4.2na.4h2o,acmc-20ajol,disodium chromate tetrahydrate,ksc492q3d,disodium chromate 2-tetrahydrate,sodium chromate tetrahydrate, reagent grade,sodium chromate tetrahydrate, p.a
22146515
disodium;dioxido(dioxo)chromium;tetrahydrate
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
10034-82-9
CrH8Na2O8
234.032
MFCD00149167
AVHUVTPOXZEIRG-UHFFFAOYSA-N
sodium chromate tetrahydrate,cro4.2na.4h2o,acmc-20ajol,disodium chromate tetrahydrate,ksc492q3d,disodium chromate 2-tetrahydrate,sodium chromate tetrahydrate, reagent grade,sodium chromate tetrahydrate, p.a
22146515
disodium;dioxido(dioxo)chromium;tetrahydrate
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
Applications
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.
UN Number UN3288
Merck Index 14,8601
Solubility Information Soluble in water.
Formula Weight 234.04 (161.97 Anhydrous)
Percent Purity ≥99%
Density 2.73
Odor Odorless
Quantity 100g
Chemical Name or Material Sodium chromate tetrahydrate

GHS H Statement
H301-H330-H334-H340-H350-H360-H372-H314-H318-H312-H317
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H312-H314-H317-H330-H334-H335-H340-H350-H360-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-889-5

TSCA : Yes

Recommended Storage : Ambient temperatures