Learn More
Sodium Citrate, 0.5 M Buffer Solution, pH 5.0, PROMO, Thermo Scientific Chemicals
Description
Applications
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.
Solubility
It is soluble in water (29.4 g/L) at 20°C. Insoluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Specifications
Specifications
Chemical Name or Material | Sodium citrate buffer solution |
Color | Colorless |
Concentration | 0.5 M |
pH | 5.0 |
Physical Form | Liquid |
Quantity | 500 mL |
MDL Number | MFCD00012462 |
Solubility Information | Soluble in water at 20°C. |
RUO - Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.