Learn More
Sodium Citrate, 0.5 M Buffer Solution, pH 5.5, PROMO, Thermo Scientific Chemicals
Description
Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application. It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts.
Solubility
Soluble in water.
Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.

Specifications
Specifications
Chemical Name or Material | Sodium citrate buffer solution |
Color | Colorless |
Concentration | 0.5 M |
pH | 5.5 |
Physical Form | Liquid |
Quantity | 500 mL |
MDL Number | MFCD00012462 |
Solubility Information | Soluble in water. |
RUO - Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.