Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
Description
Applications
Sodium citrate, 0.5 M buffer solution, pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
Solubility
Fully miscible in water.
Notes
Incompatible with heat and oxidizing agents.

Specifications
Specifications
Chemical Name or Material | Sodium citrate buffer solution |
Color | Colorless |
Concentration | 0.5 M |
pH | 6.0 |
Physical Form | Liquid |
Quantity | 500 mL |
MDL Number | MFCD00012462 |
Solubility Information | Fully miscible in water. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction