Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sodium Cromoglicate
GREENER_CHOICESDP

Catalog No. 642450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 64-245-0 Supplier Tocris Bioscience™ Supplier No. 6424/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Promotes ESCs/iPSCs differentiation into pancreatic endocrine islet cells

Sodium Cromoglicate is a promotes transformation ESCs/iPSCs into pancreatic endocrine islet cells. Facilitates differentiation from endocrine precursors. Also inhibits mast cell degranulation and histamine release. Antiasthmatic.

Specifications

Chemical Name or Material 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid] disodium salt
CAS 15826-37-6
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C23H14Na2O11
Formula Weight Observed MW: 512.33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.