Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals

Catalog No. AA1213736
Click to view available options
Quantity:
100 g
500 g
2 kg

Sodium cyanide is used as a starting material for the preparation of Reissert compounds, cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, in electroplating and for extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point ca 1500°C
Melting Point 563°C
Color Gray
Physical Form Granules
Quantity 500 g
Assay Percent Range 95% min.
Beilstein 3587243
Solubility Information Soluble in water,ammonia,methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.
Formula Weight 49.01
Grade ACS
Odor Weak, characteristic
Sensitivity Hygroscopic
Appearance White powder
Density 1.61 g/mL
Chemical Name or Material Sodium cyanide, For analysis ACS
Show More Show Less
Hazard Category H290-H300+H310+H330-H372
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P390-P501c
DOTInformation DOT Class: 6.1, Packaging Group: I
EINECSNumber 205-599-4
RTECSNumber VZ7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.