Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cyanide, Reagent, ACS, 95%, Spectrum™ Chemical
SDP

Catalog No. 18606471 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle

S1260, 143-33-9, NaCN

Spectrum™ Chemical Sodium Cyanide, Reagent, ACS is a solid compound that is inorganic, water-soluble and white. A highly toxic salt, it is mainly used in gold mining because cyanide has a high affinity and reactivity with metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade ACS
Packaging Poly Bottle
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.