Learn More
Sodium cyanoborohydride, 95%
Description
Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300°C (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 95% |
| Solubility Information | Soluble in water,THF,methanolSoluble in water,diglyme,tetrahydrofuran and methanol. Slightly soluble in dimethyl sulfoxide. Insoluble in diethyl ether. |
| IUPAC Name | sodium;cyanoboron(1-) |
| Formula Weight | 62.84 |
| Sensitivity | Moisture sensitive |
| Density | 1.20 g/cm3 at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H228-H261-H300+H310+H330-H314 |
| Precautionary Statement | P210-P223-P231+P232-P240-P241-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 4.3, Packaging Group: II |
| EINECSNumber | 247-317-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.