Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanoborohydride, 95%

Catalog No. AA8783914
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AA8783914

Supplier: Thermo Scientific Chemicals 08783914

Only null left
Add to Cart

Selective Reducing Agent For Aldehydes, Ketones, Oximes | CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol

Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive; Highly flammable
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25895-60-7
Molecular Formula CH3BNNa
Molecular Weight (g/mol) 62.84
MDL Number MFCD00003516
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
PubChem CID 20587905
SMILES [Na+].[BH3-]C#N

Specifications

Melting Point >300°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 95%
Solubility Information Soluble in water,THF,methanolSoluble in water,diglyme,tetrahydrofuran and methanol. Slightly soluble in dimethyl sulfoxide. Insoluble in diethyl ether.
IUPAC Name sodium;cyanoboron(1-)
Formula Weight 62.84
Sensitivity Moisture sensitive
Density 1.20 g/cm3 at 20°C
Chemical Name or Material Sodium cyanoborohydride
Show More Show Less
Hazard Category H228-H261-H300+H310+H330-H314
Precautionary Statement P210-P223-P231+P232-P240-P241-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 247-317-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.