Learn More
Sodium cyclamate, 98%
Description
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose's. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose′s. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.
Solubility
Soluble in water (>=10 g/100 mL at 20°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | >300°C |
Quantity | 5000 g |
Beilstein | 4166868 |
Solubility Information | Soluble in water (>=10g/100 mL at 20°C). |
IUPAC Name | sodium;N-cyclohexylsulfamate |
Formula Weight | 201.22 |
Percent Purity | 98% |
Chemical Name or Material | Sodium cyclamate |
Safety and Handling
Hazard Category | H302 |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
EINECSNumber | 205-348-9 |
RTECSNumber | GV7350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.