Alfa Aesar™ Sodium cyclopentadienide, 2-3M in THF

Reagent for the preparation of metallocenes

Manufacturer: Alfa Aesar™ 88300JT

Quantity 0.5 mole

 View more versions of this product

Catalog No. AA88300JT

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4984-82-1
    Molecular Formula C5H5Na
    Formula Weight 88.09
    MDL Number MFCD00012161
    UN Number UN3399
    Chemical Name or Material Sodium cyclopentadienide

    Specifications

    Formula Weight 88.09
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H261-H314-H351-H335
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.
    TSCA TSCA
    Density 0.938g/mL
    CAS 4984-82-1
    UN Number UN3399
    Concentration 2 to 3M in THF
    Quantity 0.5 mole
    Physical Form Liquid
    Health Hazard 1 Danger
    Flash Point −17°C (1°F)
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Molecular Formula C5H5Na
    MDL Number MFCD00012161
    Color Pink to red
    Odor Ether-like
    Sensitivity Air & Moisture Sensitive
    Chemical Name or Material Sodium cyclopentadienide

    GHS H Statement
    H225-H261-H314-H351-H335
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit