Learn More
Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals
Description
Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium Deoxycholate Monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 0.98 |
| Beilstein | 3643164 |
| Solubility Information | Soluble in water. |
| IUPAC Name | (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid;sodium;hydrate |
| Formula Weight | 432.58 (414.58 Anhydrous) |
| Chemical Name or Material | Sodium Deoxycholate Monohydrate |
Safety and Handling
| Hazard Category | H302-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| EINECSNumber | 206-132-7 |
| RTECSNumber | FZ2250000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.