Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium dichloroacetate, 97%

Click to view available options
Quantity:
10g
50g
250g
2156-56-1
C2HCl2NaO2
150.918
MFCD00070489
LUPNKHXLFSSUGS-UHFFFAOYSA-M
sodium dichloroacetate,sodium 2,2-dichloroacetate,dichloroacetic acid sodium salt,acetic acid, dichloro-, sodium salt,ceresine,dichloroacetate sodium,dichloroctan sodny czech,sodium dichloroacetate usan,ccris 7697,dichloroacetic acid, sodium salt
517326
sodium;2,2-dichloroacetate
C(C(=O)[O-])(Cl)Cl.[Na+]
2156-56-1
C2HCl2NaO2
150.918
MFCD00070489
LUPNKHXLFSSUGS-UHFFFAOYSA-M
sodium dichloroacetate,sodium 2,2-dichloroacetate,dichloroacetic acid sodium salt,acetic acid, dichloro-, sodium salt,ceresine,dichloroacetate sodium,dichloroctan sodny czech,sodium dichloroacetate usan,ccris 7697,dichloroacetic acid, sodium salt
517326
sodium;2,2-dichloroacetate
C(C(=O)[O-])(Cl)Cl.[Na+]
Applications
Sodium dichloroacetate finds medicinal application in removing warts and other skin growths. It is used as an antidote for lactic acidosis and in the treatment of stroke. It is studied as potential drugs, since it inhibits the enzyme pyruvate dehydrogenase kinase.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Melting Point 197°C to 199°C (decomposition)
Quantity 10g
Formula Weight 150.93
Percent Purity 97%
Chemical Name or Material Sodium dichloroacetate

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning