Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dichromate Dihydrate, ACS Reagent, Reagents

Catalog No. p-200020983
Click to view available options
Quantity:
100 g
500 g

Sodium Dichromate Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 298.00
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym Sodium Bichromate
IUPAC Name disodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
SMILES O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Boiling Point 400°C
Melting Point 357°C
Color Orange/Red
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.35
Chemical Name or Material Sodium Dichromate Dihydrate, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.