Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Dichromate Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Specifications
Specifications
Boiling Point | 400°C |
Melting Point | 357°C |
Color | Orange |
pH | 3.5 to 3.9 |
Physical Form | Solid |
Quantity | 500 g |
Assay Percent Range | 99.5 to 100.5 % |
UN Number | 1479 |
Formula Weight | 298 |
Percent Purity | 99.5 to 100.5% |
Show More |
Safety and Handling

Hazard Category | Signal Word: DANGER! |
Precautionary Statement | Emergency Overview Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin. |
DOTInformation | DOT Class 5.1, : Oxidizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction