Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dichromate Dihydrate, Honeywell Fluka™

Catalog No. p-7071779
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 kg
Linear Formula Na2Cr2O7 · 2H2O
UN Number 3086
Formula Weight 298g/mol
Percent Purity ≥99%
Grade purum
Chemical Name or Material Sodium Dichromate Dihydrate
Hazard Statement P201-P210-P260-P284-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P313-P403 + P233
Precautionary Statement H272-H301-H312-H314-H317-H330-H334-H340-H350-H360FD-H372-H410
EINECSNumber 234-190-3
RTECSNumber HX7750000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.