Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dichromate dihydrate, Reagent Grade

Catalog No. p-4824949
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 7789-12-0 | Cr2H4Na2O9 | 297.995 g/mol

Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitors in undercoats and primers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 100°C-2H2O
Color Red
Physical Form Crystalline
Quantity 100 g
Solubility Information Soluble in waterSoluble in water. Insoluble in alcohol.
Formula Weight 298.00 (261.97 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Appearance Red crystals
Density 2.348
Chemical Name or Material Sodium dichromate dihydrate
Show More Show Less
Hazard Category H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 234-190-3
RTECSNumber HX7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.