Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium dicyanamide, 97%, pure
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300°C |
| Color | Pink or White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 2% max. (NaCl) (Argentometry) |
| Packaging | Plastic bottle |
| Linear Formula | NaN(CN)2 |
| Quantity | 100 g |
| Beilstein | 03, III, 154 |
| Solubility Information | Solubility in water: 260g/L (30°C). Other solubilities: insoluble in aceton,etoh and ethyl acetate |
| IUPAC Name | sodium;dicyanoazanide |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lens |
| EINECSNumber | 217-703-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction