Learn More
Sodium dihydrogen citrate, 99%
Description
Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Hygroscopic. Incompatible with bases, reducing agents and oxidizing agents.
Specifications
Specifications
| Melting Point | 211°C to 212°C |
| Quantity | 2000 g |
| Beilstein | 3920246 |
| Solubility Information | Soluble in water. Insoluble in ethanol. |
| Formula Weight | 214.11 |
| Percent Purity | 99% |
| Chemical Name or Material | Sodium dihydrogen citrate |
Safety and Handling
| Hazard Category | H319-H500 |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 242-734-6 |
| RTECSNumber | GE7950000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.