Learn More
Sodium dihydrogen phosphate, 96%
Description
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It was applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 96% |
Solubility Information | Soluble in water. Insoluble in alcohol,ether and chloroform. |
Formula Weight | 119.98 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White Powder |
Chemical Name or Material | Sodium dihydrogen phosphate |
Safety and Handling
Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
EINECSNumber | 231-449-2 |
RTECSNumber | WA1900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.