Learn More
Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis)
Description
Sodium dihydrogen phosphate monohydrate is used as a food additive. It finds use as mordant in dyeing. It is also used in the manufacture of enamels, chemicals, pharmaceuticals, ceramics, detergents, and boiler compounds. In addition, it is used as a fireproofing agent, thickening agent, emulsifier, and as a reagent and buffer in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C to H2O |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.998% (metals basis) |
| Solubility Information | Soluble in water; insoluble in ethanol,ether and chloroform. |
| Formula Weight | 137.99 (119.98 Anhydrous) |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | White powder |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. |
| EINECSNumber | 231-449-2 |
| RTECSNumber | WA1900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.