Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium DL-Malate, MP Biomedicals

Catalog No. ICN218050 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 676-46-0
Molecular Formula C4H4Na2O5
Molecular Weight (g/mol) 178.051
InChI Key WPUMTJGUQUYPIV-UHFFFAOYSA-L
Synonym disodium malate, sodium malate, disodium dl-malate, dl-malic acid disodium salt, disodium 2-hydroxybutanedioate, natriummalat german, sodium dl-malate, butanedioic acid, hydroxy-, disodium salt, malic acid, disodium salt, sodium dl-maleate
PubChem CID 8736
IUPAC Name disodium;2-hydroxybutanedioate
SMILES C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]

Specifications

Quantity 10 g
Formula Weight 178.05
Chemical Name or Material Sodium DL-Malate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.