Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, 95%, BAKER™, J.T. Baker™

Catalog No. 02004086 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-086 1 kg Poly Bottle
02-004-085 500 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-004-086 Supplier Avantor Supplier No. L05002
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 1 kg
Flash Point 170.00°C
Packaging Poly Bottle
Specific Gravity 1L = 0.40kg
Formula Weight 288.38
Percent Purity 95%
Chemical Name or Material Sodium Dodecyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.