Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Dodecyl Sulfate, 95%, BAKER™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Specifications
| Quantity | 500 g |
| Flash Point | 170.00°C |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 0.40kg |
| Formula Weight | 288.38 |
| Percent Purity | 95% |
| Chemical Name or Material | Sodium Dodecyl Sulfate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction