Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl, 1 kg PROMO
Click to view available options
:
Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl
Description
This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).
Highlights:
- Popular anionic detergent for a variety of protein methods
- Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
- Common component of cell lysis buffers
Detergent properties (pure C12):
- Molecular Weight: 288.5
- Detergent Class: Ionic (anionic)
- Aggregation Number: 62
- Micelle Molecular Weight: 18,000
- Critical Micelle Concentration (CMC): 6 to 8mM (0.1728 to 0.2304%, w/v)
- Cloud Point: greater than 100°C
- Dialyzable: No
Storage Requirements
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
- Should be stored between the following temperatures: 20°C to 25°C
Specifications
Specifications
Color | White |
Product Type | Detergent |
Solubility | 10% (aq, w/v) solution must be clear, colorless to slightly yellow |
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C. |
Form | Powder |
Reagent Type | Detergent (Pure) |
Quantity | 1 kg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction