Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02004082
Click to view available options
Quantity:
100 g
1 kg
2 kg
Packaging:
Poly Pail
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Electrophoresis and Protein Solubilization; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym SDS
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 1 kg
Flash Point 170.00°C
Packaging Poly bottle
Specific Gravity 1L = 0.40kg
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Grade Ultrapure Bioreagent
Chemical Name or Material Sodium Dodecyl Sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.