Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dodecylbenzenesulfonate (hard type) (mixture) 95.0+%, TCI America™
SDP

Catalog No. D099025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D099025G 25 g
D0990500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D099025G Supplier TCI America Supplier No. D099025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
MDL Number MFCD00011508
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Synonym Alkylbenzenesulfonic Acid Sodium Salt, Dodecylbenzenesulfonic Acid Sodium Salt, Laurylbenzenesulfonic Acid Sodium Salt, Sodium Alkylbenzenesulfonate, Sodium Laurylbenzenesulfonate

Specifications

Melting Point 204°C
Color White-Yellow
pH 5.5 - 8.0 (10g/L)
Quantity 25 g
UN Number 3077
Formula Weight 348.48
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dodecylbenzenesulfonate (hard type) (mixture)
EINECSNumber (3)-1906&(3)-1949
RTECSNumber DB6825000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.