Learn More
Sodium ethoxide, 21% w/w in ethanol
Description
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.
Solubility
Miscible with alcohol.
Notes
Moisture sensitive. It hydrolyzes rapidly into sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.

Specifications
Specifications
Density | 0.868 |
Flash Point | 20°C (68°F) |
Refractive Index | 1.3875 |
Linear Formula | NaOCH2CH3 |
Quantity | 2500 mL |
UN Number | UN3274 |
Beilstein | 3593646 |
Sensitivity | Air and moisture sensitive |
Merck Index | 14,8613 |
Solubility Information | Miscible with alcohol. |
Show More |
Safety and Handling
Hazard Category | H226-H314-H335 |
Hazard Statement | GHS H Statement H225-H314 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
Precautionary Statement | P201-P202-P210-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S. |
EINECSNumber | 205-487-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
For Manufacturing and Laboratory Use. Not for beverage use
Your input is important to us. Please complete this form to provide feedback related to the content on this product.