Learn More
Sodium ethoxide, 21% w/w soln. in ethanol, packaged under Argon in resealable ChemSeal™ bottles
Description
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon

Specifications
Specifications
Density | 0.868 g/mL |
Flash Point | 20°C (68°F) |
Packaging | ChemSeal™ bottles |
Refractive Index | 1.385 |
Linear Formula | NaOCH2CH3 |
Quantity | 500 mL |
UN Number | UN3274 |
Beilstein | 3593646 |
Sensitivity | Moisture sensitive |
Merck Index | 14,8613 |
Show More |
Safety and Handling
Hazard Category | H226-H314-H335 |
Hazard Statement | GHS H Statement H224-H314-H318 Extremely flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P201-P202-P210-P233-P235-P235+P410-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S. |
EINECSNumber | 205-487-5 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
For Manufacturing and Laboratory Use. Not for beverage use
Your input is important to us. Please complete this form to provide feedback related to the content on this product.