Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium ethoxide, pure, 21% in ethanol
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow to Brown |
| Boiling Point | 91.0°C |
| Linear Formula | C2H5ONa |
| Beilstein | 01, 311 |
| Fieser | 01,1065; 03,265; 04,451; 06,540 |
| Density | 0.8680g/mL |
| Merck Index | 15, 8749 |
| Solubility Information | Solubility in water: miscible (hydrolyse). |
| Formula Weight | 68.04 |
| Percent Purity | 20.5 to 21.5% (NaOC2H5) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Highly flammable liquid and vapor. Reacts violently with water. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 205-487-5 |
| TSCA | TSCA |
| Recommended Storage | May darken on storage with no change in quality |
For Manufacturing and Laboratory Use. Not for beverage use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction