Learn More
Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Sodium fluoride is used in laundry souring, manufacturing enamels, manufacturing coated papers, wood preservative , foam production, opal glass, ore flotation and water fluoridation. It is also acts as an inhibitor of serine/threonine phosphatases widely used in bone and dental research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1704°C |
| Melting Point | 993°C |
| Color | White |
| Physical Form | Pieces |
| Quantity | 10 g |
| Linear Formula | NaF |
| UN Number | UN1690 |
| Merck Index | 14,8618 |
| Solubility Information | It is soluble in water,slightly soluble in HF,ammonia. Negligible in alcohol,acetone,SO2,dimethylformamide. |
| Formula Weight | 41.99 |
| Show More |
Safety and Handling
| Hazard Category | H301-H315-H319 |
| Hazard Statement | GHS H Statement H301-H315-H319 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM FLUORIDE, SOLID |
| EINECSNumber | 231-667-8 |
| RTECSNumber | WB0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.