Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA1100322
Click to view available options
Quantity:
5 g
25 g
100 g

It is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including its use in synthetic chemistry, for example in Finkelstein reaction for preparing fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium;fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 993°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 41.99
Odor Odorless
Refractive Index 1.336
Appearance White powder
Vapor Pressure 1.3 hPa (1mm Hg) at 1077°C
Density 2.78 g/cm3 at 20°C
Chemical Name or Material Sodium fluoride
Show More Show Less
Hazard Category H301-H315-H319
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 231-667-8
RTECSNumber WB0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.