Sodium fluoride, 99.995% (metals basis excluding K), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1296430

Quantity 250g

 View more versions of this product

Catalog No. AA1296430

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7681-49-4
    Physical Form Powder
    Assay Percent Range 99.9995% (metals basis excluding K)
    Molecular Formula NaF
    Formula Weight 41.99
    MDL Number MFCD00003524
    Chemical Name or Material Sodium fluoride

    Specifications

    Physical Form Powder
    Formula Weight 41.99
    Quantity 250g
    Color White
    DOT Information Hazard Class: 6.1, Packing Group: III
    EINECS Number 231-667-8
    CAS 7681-49-4
    Vapor Pressure 1.3 hPa (1mm Hg) at 20°C
    Odor Odorless
    Assay Percent Range 99.9995% (metals basis excluding K)
    Molecular Formula NaF
    Appearance White powder
    MDL Number MFCD00003524
    Health Hazard 1 H301-H315-H319: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation.
    Chemical Name or Material Sodium fluoride

    H301-H315-H319: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit