Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fluoride, 99+%, for analysis, Thermo Scientific Chemicals

Catalog No. p-41953
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1700.0°C
Melting Point 993.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99+%
Linear Formula NaF
Fieser 01,1073; 02,382
Merck Index 15, 8754
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol
Formula Weight 41.98
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.2% max. (150°C)
Packaging Plastic bottle
Chemical Name or Material Sodium fluoride, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs
EINECSNumber 231-667-8
RTECSNumber WB0350000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.