Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Fluoride, ACS Reagent, ≥99%, Solstice

Catalog No. 60045091 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-091 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-091 Supplier Solstice Supplier No. 201154500G

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 990°C
Color Colorless
pH 8 to 10 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula NaF
UN Number UN1690
IUPAC Name sodium fluoride
Formula Weight 41.99g/mol
Percent Purity ≥99%
Grade ACS Reagent
Packaging Poly Bottle
Vapor Pressure 1.4mmHg (0°C)
Density 2.7g/cu.cm
Chemical Name or Material Sodium Fluoride
Show More Show Less
Hazard Statement Missing Phrase - N15.00950417-P305 + P351 + P338
Precautionary Statement H301-H315-H319
DOTInformation 6.1/PGIII
EINECSNumber 231-667-8
RTECSNumber WB0350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.