Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Fluoride, Purified, Reagents

Catalog No. 01337018
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 lb.

Sodium Fluoride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 990°C
Color White
Physical Form Solid
Quantity 50 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.78
Chemical Name or Material Sodium Fluoride, Purified
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.