Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium formaldehydesulfoxylate dihydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
6035-47-8
CH3NaO32H2O
154.11
MFCD00150598
UCWBKJOCRGQBNW-UHFFFAOYSA-M
sodium hydroxymethanesulfinate dihydrate,sodium formaldehydesulfoxylate dihydrate,methanesulfinic acid, hydroxy-, monosodium salt, dihydrate,sodium formaldehyde sulfoxylate,sodium dihydrate hydroxymethanesulfinate,sodium formaldehyde sulfoxylate nf,formaldehyde sodium sulfoxylate dihydrate,monosodium hydroxymethanesulfinate dihydrate,79-25-4 parent,hydroxymethanesulfinic acid monosodium salt dihydrate
23666330
sodium;hydroxymethanesulfinate;dihydrate
C(O)S(=O)[O-].O.O.[Na+]
6035-47-8
CH3NaO32H2O
154.11
MFCD00150598
UCWBKJOCRGQBNW-UHFFFAOYSA-M
sodium hydroxymethanesulfinate dihydrate,sodium formaldehydesulfoxylate dihydrate,methanesulfinic acid, hydroxy-, monosodium salt, dihydrate,sodium formaldehyde sulfoxylate,sodium dihydrate hydroxymethanesulfinate,sodium formaldehyde sulfoxylate nf,formaldehyde sodium sulfoxylate dihydrate,monosodium hydroxymethanesulfinate dihydrate,79-25-4 parent,hydroxymethanesulfinic acid monosodium salt dihydrate
23666330
sodium;hydroxymethanesulfinate;dihydrate
C(O)S(=O)[O-].O.O.[Na+]
Reduction reagent
Melting Point 63.0°C to 64.0°C
Packaging Fibre drum
Quantity 25kg
Assay Percent Range 98%
Linear Formula HOCH2SO2Na·2H2O
Merck Index 15,8755
Solubility Information Solubility in water: 1088g/L. Other solubilities: practically insoluble in abs.alcohol, ether and, benzene
Formula Weight 154.11
Percent Purity 98%
Chemical Name or Material Sodium formaldehydesulfoxylate dihydrate

GHS H Statement:
Suspected of causing genetic defects.
Contact with acids liberates toxic gas.

GHS P Statement:
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning