Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fumarate, 98%

Catalog No. AAA1127636
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 17013-01-3 | C4H4Na2O4 | 162.052 g/mol

Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17013-01-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00064567
InChI Key VXXVUHAXJHEYFH-SEPHDYHBSA-N
Synonym disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
PubChem CID 87110070
IUPAC Name (E)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Assay Percent Range 98%
Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.04
Percent Purity 98%
Chemical Name or Material Sodium fumarate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-087-7
RTECSNumber LT1830000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.