Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fumarate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
17013-01-3
C4H2Na2O4
160.04
MFCD00064567
VXXVUHAXJHEYFH-SEPHDYHBSA-N
disodium fumarate,sodium fumarate,sodium fumarate dibasic,fumaric acid, disodium salt,fumaric acid disodium salt,fumaran sodny czech,fumaran sodny,disodium butenedioate,di-sodium fumarate,sodium monofumarate
87110070
(E)-but-2-enedioic acid;sodium
C(=CC(=O)O)C(=O)O.[Na].[Na]
17013-01-3
C4H2Na2O4
160.04
MFCD00064567
VXXVUHAXJHEYFH-SEPHDYHBSA-N
disodium fumarate,sodium fumarate,sodium fumarate dibasic,fumaric acid, disodium salt,fumaric acid disodium salt,fumaran sodny czech,fumaran sodny,disodium butenedioate,di-sodium fumarate,sodium monofumarate
87110070
(E)-but-2-enedioic acid;sodium
C(=CC(=O)O)C(=O)O.[Na].[Na]
Infrared Spectrum Authentic
Quantity 5g
Color White
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula NaO2CCH=CHCO2Na
Beilstein 02,74
Merck Index 10,4165
Solubility Information Solubility in water: 228g/L water. Other solubilities: insoluble in ethanol, aceton
Formula Weight 160.04
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Sodium fumarate
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

EINECSNumber : 241-087-7