Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium galacturonate anhydrous, 98%

Catalog No. p-7043268
Click to view available options
Quantity:
5 g
25 g

CAS: 14984-39-5 | C6H9NaO7 | 216.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14984-39-5
Molecular Formula C6H9NaO7
Molecular Weight (g/mol) 216.121
MDL Number MFCD06201013
InChI Key MSXHSNHNTORCAW-NSBQNPTMSA-M
Synonym d-+-galacturonic acid sodium salt
PubChem CID 122174033
IUPAC Name sodium;(4S)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES C1(C(C(OC(C1O)O)C(=O)[O-])O)O.[Na+]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Formula Weight 216.12
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium galacturonate, anhydrous
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.