Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium glycocholate hydrate, 98%
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Sodium glycocholate hydrate is a conjugated bile salt
- It is an ionic detergent
Applications
- Sodium glycocholate hydrate is used to determine serum total bile acids by fluorimetric and enzymatic methods.
- It has also been used in in vitro studies to investigate its effect on micelle formation and solubilization of testosterone.
- This compound is considered a detergent used to solubilize fats for absorption.
- This chemical product is synthetically derived from a natural compound, cholic acid.
- Sodium glycocholate hydrate has been tested in the laboratory for its ability to form a liposomal delivery system to enhance oral insulin delivery. The in vitro results demonstrated that sodium glycocholate liposomes showed promising activity and can be useful to create a new approach to orally deliver insulin more efficiently.
Specifications
Specifications
| Color | Colorless to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.98 |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Specific Rotation Condition | + 29.50 (20.00°C c=1,H2O) |
| Specific Rotation | +28° to +31° (20°C, 589nm) (c=1, H2O) on dry basis |
| Formula Weight | 487.61 |
| Percent Purity | ≥97.5% (on dry basis) (Total base) |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Skin corrosion/irritation (category 2) |
| Hazard Statement | Causes serious eye irritation, Causes skin irritation |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction