Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexachloroplatinate(IV) hexahydrate

Catalog No. p-4824950
Click to view available options
Quantity:
1 g
5 g

For Use As a Catalyst | CAS: 19583-77-8 | Cl6H12Na2O6Pt | 561.85 g/mol

Catalyst.Sodium hexachloroplatinate(IV) hexahydrate is used as a catalyst. It acts as a source of platinum in the preparation of platinum complex. It is also used in the synthesis of platinum nanoparticles using glutathione as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19583-77-8
Molecular Formula Cl6H12Na2O6Pt
Molecular Weight (g/mol) 561.85
MDL Number MFCD00149172
InChI Key XGFSWVNCHVSCSP-UHFFFAOYSA-H
Synonym sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
PubChem CID 24846071
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 110°C-6H2O
Color Orange
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water,alcohol. Insoluble in ether.
IUPAC Name disodium;hexachloroplatinum(2-);hexahydrate
Formula Weight 561.88 (453.79 Anhydrous)
Odor Odorless
Appearance Orange powder
Density 2.5
Chemical Name or Material Sodium hexachloroplatinate(IV) hexahydrate
Show More Show Less
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 240-983-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.