Learn More
Sodium hexafluoroaluminate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Sodium aluminum hexafluoride is used as a solvent (or flux) for electrolysis aluminum oxides such as bauxite, whitener for enamels and an opacifier for glass and in the industrial production of aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1000°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Soluble in water(0.4g/L). |
Formula Weight | 209.95 |
Odor | Odorless |
Density | 2.9 g/cm3 at 20°C |
Chemical Name or Material | Sodium hexafluoroaluminate |
Safety and Handling
Hazard Category | H332-H372 |
Hazard Statement | Do not breathe dust. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets. |
Precautionary Statement | P260-P264b-P270-P271-P304+P340-P312-P314-P501c |
DOTInformation | DOT Class: 9; Packaging Group: III |
EINECSNumber | 237-410-6 |
RTECSNumber | BD0075000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.