Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)

Catalog No. AA1262006
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA1262006

Supplier: Thermo Scientific Chemicals 01262006

Only null left
Add to Cart

CAS: 12325-31-4 | H6Na2O6Pt | 343.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12325-31-4
Molecular Formula H6Na2O6Pt
Molecular Weight (g/mol) 343.11
MDL Number MFCD00014236
InChI Key LDMNYTKHBHFXNG-UHFFFAOYSA-H
IUPAC Name platinum(4+) disodium hexahydroxide
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Pt+4]

Specifications

Melting Point 150°C (decomposition)
Color Yellow to Brown
Linear Formula Na2Pt(OH)6
Quantity 5 g
Formula Weight 343.1
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Sodium hexahydroxyplatinate(IV)
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 235-590-0
RTECSNumber TP1756000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.